Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006230773.2 | XP_006230835.1 | 10 - 11 | 1472 | 83 | |
XM_006230774.2 | XP_006230836.1 | 10 - 11 | 1437 | 83 | ||
GenBank mRNA | BC061558.1 | - | 9 - 10 | 1951 | 6919414 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |