Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760054.1 | XP_008758276.1 | 1 - 2 | 964 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Ion Channel Rattus norvegicus Probe spans exons |