Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsdmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130553.1 | NP_001124025.1 | 7 - 8 | 920 | 96 | |
XM_006241809.2 | XP_006241871.1 | 7 - 8 | 1575 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |