Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133526.1 | NP_598210.1 | 6 - 7 | 798 | 131 | |
GenBank mRNA | BC061785.1 | - | 6 - 7 | 654 | 5623788 | 131 |
FQ214867.1 | - | 6 - 7 | 706 | 131 | ||
FQ215876.1 | - | 2 - 3 | 170 | 131 | ||
FQ216712.1 | - | 6 - 7 | 827 | 131 | ||
FQ220073.1 | - | 2 - 3 | 170 | 131 | ||
FQ220093.1 | - | 2 - 3 | 170 | 131 | ||
FQ227464.1 | - | 6 - 7 | 815 | 131 | ||
FQ229225.1 | - | 2 - 3 | 170 | 131 | ||
FQ229522.1 | - | 6 - 7 | 716 | 131 | ||
Y13275.2 | - | 6 - 7 | 798 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |