Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam203a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007707.1 | NP_001007708.1 | 2 - 3 | 791 | 176 | |
GenBank mRNA | BC079158.1 | - | 2 - 3 | 791 | 7129210 | 176 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |