Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam203a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007707.1 | NP_001007708.1 | 4 - 5 | 979 | 127 | |
GenBank mRNA | BC079158.1 | - | 4 - 5 | 979 | 7129210 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |