Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Top1mt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002798.1 | NP_001002798.1 | 8 - 9 | 1163 | 117 | |
XM_006241784.2 | XP_006241846.1 | 8 - 9 | 1363 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |