Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031122.1 | NP_112384.1 | 3 - 4 | 305 | 89 | |
GenBank mRNA | BC078804.1 | - | 3 - 4 | 312 | 7109473 | 89 |
FQ228746.1 | - | 3 - 4 | 267 | 89 | ||
X82021.1 | - | 3 - 4 | 305 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |