Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011992.1 | NP_001011992.1 | 1 - 2 | 65 | 72 | |
XM_006241602.2 | XP_006241664.1 | 1 - 2 | 169 | 72 | ||
GenBank mRNA | BC089961.1 | - | 1 - 2 | 65 | 7314200 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |