Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077669.1 | NP_001071137.1 | 7 - 8 | 1420 | 110 | |
XM_008765563.1 | XP_008763785.1 | 8 - 9 | 3597 | 110 | ||
XM_008765564.1 | XP_008763786.1 | 8 - 9 | 3475 | 110 | ||
GenBank mRNA | BC127514.1 | - | 7 - 8 | 1420 | 7100017 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |