Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ago2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021597.1 | NP_067608.1 | 1 - 2 | 222 | 150 | |
GenBank mRNA | AF195534.1 | - | 1 - 2 | 222 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |