Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025701.1 | NP_001020872.1 | 4 - 5 | 573 | 93 | |
GenBank mRNA | BC099200.1 | - | 4 - 5 | 573 | 7384021 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |