Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079894.1 | NP_001073363.1 | 6 - 7 | 591 | 101 | |
XM_008759909.1 | XP_008758131.1 | 6 - 7 | 638 | 101 | ||
GenBank mRNA | BC128752.1 | - | 6 - 7 | 591 | 7100072 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |