Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006230316.2 | XP_006230378.1 | 7 - 8 | 1402 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |