Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem41b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012358.1 | NP_001012358.1 | 1 - 2 | 244 | 78 | |
XM_008759766.1 | XP_008757988.1 | 1 - 2 | 365 | 78 | ||
GenBank mRNA | BC089866.1 | - | 1 - 2 | 244 | 7321590 | 78 |
FQ228941.1 | - | 1 - 2 | 305 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |