Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765260.1 | XP_008763482.1 | 1 - 2 | 588 | 56 | |
XM_008765261.1 | XP_008763483.1 | 1 - 2 | 587 | 56 | ||
GenBank mRNA | BC083815.1 | - | 1 - 2 | 108 | 7110420 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Nuclear Receptor Mammalian Gene Collection Rattus norvegicus Probe spans exons |