Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759807.1 | XP_008758029.1 | 4 - 5 | 1435 | 89 | |
XM_238899.7 | XP_238899.4 | 3 - 4 | 1238 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Protease Rattus norvegicus Probe spans exons |