Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdfi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109301.1 | NP_001102771.1 | 3 - 4 | 560 | 124 | |
XM_006244475.2 | XP_006244537.1 | 3 - 4 | 321 | 124 | ||
XM_006244476.2 | XP_006244538.1 | 3 - 4 | 514 | 124 | ||
XM_008766877.1 | XP_008765099.1 | 3 - 4 | 487 | 124 | ||
GenBank mRNA | BC158727.1 | - | 2 - 3 | 482 | 9091612 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |