Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106500.1 | NP_001099970.1 | 4 - 5 | 1108 | 132 | |
XM_006234737.1 | XP_006234799.1 | 4 - 5 | 988 | 132 | ||
XM_006234738.2 | XP_006234800.1 | 5 - 6 | 985 | 132 | ||
XM_006234739.2 | XP_006234801.1 | 4 - 5 | 976 | 132 | ||
XM_006234740.2 | XP_006234802.1 | 4 - 5 | 1232 | 132 | ||
XM_006234741.2 | XP_006234803.1 | 4 - 5 | 1132 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |