Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Astl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106504.1 | NP_001099974.1 | 3 - 4 | 299 | 94 | |
XM_008762157.1 | XP_008760379.1 | 3 - 4 | 242 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Protease Rattus norvegicus Probe spans exons |