Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creb3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005562.1 | NP_001005562.1 | 2 - 3 | 730 | 64 | |
XM_006234584.1 | XP_006234646.1 | 2 - 3 | 786 | 64 | ||
GenBank mRNA | BC081951.1 | - | 2 - 3 | 730 | 7122657 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |