Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tas1r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271266.1 | NP_001258195.1 | 5 - 6 | 1603 | 56 | |
GenBank mRNA | AF127390.1 | - | 5 - 6 | 1603 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons |