Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012105.1 | NP_001012105.1 | 3 - 4 | 459 | 74 | |
XM_006239303.1 | XP_006239365.1 | 3 - 4 | 454 | 74 | ||
XM_008764253.1 | XP_008762475.1 | 2 - 3 | 365 | 74 | ||
GenBank mRNA | BC083577.1 | - | 3 - 4 | 459 | 7113925 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |