Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077648.1 | NP_001071116.1 | 2 - 3 | 162 | 122 | |
GenBank mRNA | U17837.1 | - | 2 - 3 | 162 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |