Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764746.1 | XP_008762968.1 | 2 - 3 | 274 | 64 | |
XM_008764747.1 | XP_008762969.1 | 3 - 4 | 255 | 64 | ||
XM_008764748.1 | XP_008762970.1 | 2 - 3 | 273 | 64 | ||
XM_008764750.1 | XP_008762972.1 | 2 - 3 | 273 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |