Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191818.1 | NP_001178747.1 | 6 - 7 | 622 | 68 | |
XM_008764746.1 | XP_008762968.1 | 7 - 8 | 842 | 68 | ||
XM_008764747.1 | XP_008762969.1 | 8 - 9 | 823 | 68 | ||
XM_008764748.1 | XP_008762970.1 | 7 - 8 | 841 | 68 | ||
XM_008764749.1 | XP_008762971.1 | 6 - 7 | 671 | 68 | ||
XM_008764750.1 | XP_008762972.1 | 7 - 8 | 841 | 68 | ||
XM_008764751.1 | XP_008762973.1 | 6 - 7 | 1088 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |