Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109279.1 | NP_001102749.1 | 5 - 6 | 596 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |