Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smek1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108050.2 | NP_001101520.2 | 13 - 14 | 2590 | 141 | |
NM_001271174.1 | NP_001258103.1 | 13 - 14 | 2887 | 141 | ||
GenBank mRNA | DY321166.1 | - | 4 - 5 | 627 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |