Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079709.1 | NP_001073177.1 | 2 - 3 | 1136 | 61 | |
GenBank mRNA | BC128770.1 | - | 2 - 3 | 1136 | 7127435 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons |