Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108716.1 | NP_001102186.1 | 12 - 13 | 1315 | 83 | |
XM_006240347.2 | XP_006240409.1 | 12 - 13 | 1318 | 83 | ||
XM_008764814.1 | XP_008763036.1 | 11 - 12 | 1366 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |