Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021655.2 | NP_067687.2 | 2 - 3 | 238 | 77 | |
GenBank mRNA | AF145445.1 | - | 2 - 3 | 100 | 77 | |
BC087703.1 | - | 2 - 3 | 218 | 7313842 | 77 | |
CB743841.1 | - | 2 - 3 | 294 | 77 | ||
X06832.1 | - | 2 - 3 | 223 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |