Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014043.1 | NP_001014065.1 | 2 - 3 | 882 | 120 | |
XM_006233311.2 | XP_006233373.1 | 3 - 4 | 1818 | 120 | ||
XM_008761501.1 | XP_008759723.1 | 2 - 3 | 1734 | 120 | ||
GenBank mRNA | BC085803.1 | - | 2 - 3 | 882 | 7111151 | 120 |
DQ071571.2 | - | 1 - 2 | 453 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |