Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bai2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107914.1 | NP_001101384.1 | 22 - 23 | 3450 | 169 | |
XM_006238934.1 | XP_006238996.1 | 23 - 24 | 3558 | 169 | ||
XM_006238935.2 | XP_006238997.1 | 23 - 24 | 3921 | 169 | ||
XM_006238936.1 | XP_006238998.1 | 23 - 24 | 3558 | 169 | ||
XM_006238938.1 | XP_006239000.1 | 22 - 23 | 3393 | 169 | ||
XM_006238939.1 | XP_006239001.1 | 22 - 23 | 3393 | 169 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Rattus norvegicus Probe spans exons |