Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fndc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270981.1 | NP_001257910.1 | 2 - 3 | 213 | 57 | |
GenBank mRNA | AF529211.1 | - | 1 - 2 | 119 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |