Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iffo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134703.1 | NP_001128175.1 | 6 - 7 | 875 | 66 | |
GenBank mRNA | CV105369.1 | - | 3 - 4 | 361 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |