Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Miip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017450.1 | NP_001017450.1 | 6 - 7 | 929 | 60 | |
XM_006239379.2 | XP_006239441.1 | 6 - 7 | 1004 | 60 | ||
XM_006239380.2 | XP_006239442.1 | 6 - 7 | 1102 | 60 | ||
XM_006239381.2 | XP_006239443.1 | 6 - 7 | 1031 | 60 | ||
GenBank mRNA | BC079047.1 | - | 6 - 7 | 929 | 7115212 | 60 |
FJ618907.1 | - | 6 - 7 | 1133 | 60 | ||
FQ227320.1 | - | 6 - 7 | 887 | 60 |
Gene Symbol: LOC100911456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239406.2 | XP_006239468.1 | 1004 | 60 | ||
XM_006239407.2 | XP_006239469.1 | 1102 | 60 | |||
XM_006239408.2 | XP_006239470.1 | 1128 | 60 | |||
XM_006239409.2 | XP_006239471.1 | 1031 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |