Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ece1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053596.2 | NP_446048.2 | 14 - 15 | 1685 | 74 | |
XM_006239275.2 | XP_006239337.1 | 14 - 15 | 2273 | 74 | ||
XM_006239276.2 | XP_006239338.1 | 13 - 14 | 2289 | 74 | ||
GenBank mRNA | BC072504.1 | - | 14 - 15 | 1685 | 7099038 | 74 |
D29683.1 | - | 14 - 15 | 1720 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |