Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130426.4 | NP_569110.1 | 7 - 8 | 939 | 69 | |
GenBank mRNA | AF420214.1 | - | 7 - 8 | 810 | 69 | |
AF498039.2 | - | 7 - 8 | 939 | 69 | ||
AY191268.1 | - | 939 | 69 | |||
AY191269.1 | - | 939 | 69 | |||
AY344841.1 | - | 7 - 8 | 852 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |