Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map7d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109438.1 | NP_001102908.1 | 9 - 10 | 1534 | 81 | |
XM_006238901.2 | XP_006238963.1 | 8 - 9 | 1610 | 81 | ||
XM_006238902.2 | XP_006238964.1 | 8 - 9 | 1601 | 81 | ||
XM_006238903.2 | XP_006238965.1 | 7 - 8 | 1499 | 81 | ||
XM_006238904.2 | XP_006238966.1 | 7 - 8 | 1490 | 81 | ||
XM_006238906.2 | XP_006238968.1 | 8 - 9 | 1394 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |