Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noc2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033897.1 | NP_001029069.1 | 6 - 7 | 880 | 82 | |
XM_006239584.2 | XP_006239646.1 | 7 - 8 | 794 | 82 | ||
GenBank mRNA | BC101911.1 | - | 6 - 7 | 880 | 7934070 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |