Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noc2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033897.1 | NP_001029069.1 | 8 - 9 | 1105 | 87 | |
XM_006239584.2 | XP_006239646.1 | 9 - 10 | 1019 | 87 | ||
GenBank mRNA | BC101911.1 | - | 8 - 9 | 1105 | 7934070 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |