Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmeb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109268.2 | NP_001102738.1 | 5 - 6 | 673 | 90 | |
XM_008764165.1 | XP_008762387.1 | 5 - 6 | 1006 | 90 | ||
XM_008764166.1 | XP_008762388.1 | 6 - 7 | 1369 | 90 | ||
XM_008764167.1 | XP_008762389.1 | 6 - 7 | 1366 | 90 | ||
XM_008764168.1 | XP_008762390.1 | 7 - 8 | 1257 | 90 | ||
XM_008764169.1 | XP_008762391.1 | 6 - 7 | 1200 | 90 | ||
GenBank mRNA | BC158640.1 | - | 5 - 6 | 673 | 8362091 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |