Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031770.3 | NP_113958.1 | 8 - 9 | 789 | 63 | |
XM_008766344.1 | XP_008764566.1 | 10 - 11 | 1425 | 63 | ||
GenBank mRNA | AF001953.1 | - | 8 - 9 | 789 | 63 | |
AY552803.1 | - | 8 - 9 | 789 | 63 | ||
BC089221.1 | - | 8 - 9 | 803 | 7320619 | 63 | |
DQ120491.1 | - | 8 - 9 | 796 | 63 | ||
DQ120492.1 | - | 8 - 9 | 796 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |