Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100810.1 | NP_001094280.1 | 10 - 11 | 1248 | 82 | |
XM_006233211.2 | XP_006233273.1 | 10 - 11 | 1626 | 82 | ||
XM_008761424.1 | XP_008759646.1 | 10 - 11 | 1626 | 82 | ||
XM_008761425.1 | XP_008759647.1 | 10 - 11 | 1627 | 82 | ||
GenBank mRNA | BC098742.1 | - | 8 - 9 | 926 | 82 | |
BC166571.1 | - | 10 - 11 | 1248 | 7320328 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |