Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053312.1 | NP_445764.1 | 1 - 2 | 73 | 77 | |
GenBank mRNA | BC101904.1 | - | 1 - 2 | 73 | 77 | |
BC166487.1 | - | 1 - 2 | 60 | 7103882 | 77 | |
CK356543.1 | - | 1 - 2 | 59 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |