Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053312.1 | NP_445764.1 | 6 - 7 | 793 | 68 | |
GenBank mRNA | BC101904.1 | - | 7 - 8 | 1005 | 68 | |
BC166487.1 | - | 6 - 7 | 780 | 7103882 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |