Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004227.1 | NP_001004227.1 | 5 - 6 | 484 | 110 | |
XM_008761456.1 | XP_008759678.1 | 4 - 5 | 440 | 110 | ||
GenBank mRNA | BC079374.1 | - | 5 - 6 | 484 | 7110129 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |