Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109385.2 | NP_001102855.1 | 8 - 9 | 1204 | 61 | |
GenBank mRNA | BC166815.1 | - | 8 - 9 | 1204 | 7460062 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |