Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047918.2 | NP_001041383.2 | 7 - 8 | 1207 | 101 | |
GenBank mRNA | FQ231577.1 | - | 6 - 7 | 884 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |