Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mttp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107727.1 | NP_001101197.1 | 10 - 11 | 1438 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Rattus norvegicus Probe spans exons |